2-Bromo-9-hexyl-9H-carbazole ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
B5607-1G
B5607-5G
TCB5607-1GEA
163
USD
TCB5607-1G
TCB5607-5G
2-Bromo-9-hexyl-9H-carbazole ≥98.0% (by GC)
2-Bromo-9-hexyl-9H-carbazole
Formula:
C₁₈H₂₀BrN |
MDL Number:
MFCD31618110 CAS Number: 864550-95-8 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 40.0 to 44.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...