Bis(tetrabutylammonium) dihydrogen Pyrophosphate ≥96.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
B5440-1G
B5440-200MG
TCB5440-1GEA
733
USD
TCB5440-1G
TCB5440-200MG
Bis(tetrabutylammonium) dihydrogen Pyrophosphate ≥96.0% (by titrimetric analysis)
Bis(tetrabutylammonium) dihydrogen Pyrophosphate
Formula:
C₃₂H₇₄N₂O₇P₂ Storage Temperature: Freezer |
CAS Number:
857447-79-1 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...