9-(2-Ethylhexyl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: 2-[9-(2-Ethylhexyl)-9H-carbazol-3-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 9-(2-Ethylhexyl)-9H-carbazole-3-boronic acid Pinacol ester

E1323-1G E1323-200MG
TCE1323-1GEA 230 USD
TCE1323-1G TCE1323-200MG
9-(2-Ethylhexyl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole ≥98.0% (by HPLC)
9-(2-Ethylhexyl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
Formula: C₂₆H₃₆BNO₂
MW: 405.39 g/mol
MDL Number: MFCD31618140
CAS Number: 856422-41-8

Order Now

Specification Test Results

Appearance Light yellow to Yellow clear liquid
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR