3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-(9H-Carbazol-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, (9H-Carbazol-3-yl)boronic acid Pinacol ester

T3117-1G T3117-5G
TCT3117-1GEA 34.5 USD
TCT3117-1G TCT3117-5G
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole ≥98.0% (by GC)
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
Formula: C₁₈H₂₀BNO₂
MW: 293.17 g/mol
MDL Number: MFCD16996081
CAS Number: 855738-89-5

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 194.0 to 198.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR