4,4'-Dibromo-4''-tert-butyltriphenylamine ≥95.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
N,N-Bis(4-bromophenyl)-4-tert-butylaniline
D5330-1G
D5330-5G
TCD5330-1GEA
128
USD
TCD5330-1G
TCD5330-5G
4,4'-Dibromo-4''-tert-butyltriphenylamine ≥95.0% (by HPLC, total nitrogen)
4,4'-Dibromo-4''-tert-butyltriphenylamine
Formula:
C₂₂H₂₁Br₂N |
CAS Number:
852534-22-6 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (with Total Nitrogen) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...