7-Bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carboxaldehyde ≥96.0% (by GC)
Supplier: TCI America
Synonyms:
7-Bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carbaldehyde, 2-Bromo-3,4-ethylenedioxythiophene-5-carboxaldehyde
B5396-1G
B5396-200MG
TCB5396-1GEA
321
USD
TCB5396-1G
TCB5396-200MG
7-Bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carboxaldehyde ≥96.0% (by GC)
7-Bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carboxaldehyde
Formula:
C₇H₅BrO₃S MW: 249.08 g/mol |
MDL Number:
MFCD08690306 CAS Number: 852054-42-3 |
Specification Test Results
Appearance | White to Light gray to Light yellow powder to crystal |
Purity (GC) | min. 96.0 % |
Melting point | 147.0 to 152.0 deg-C |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...