2,4-Bis(benzyloxy)-6-chloro-1,3,5-triazine ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: DBT-Cl

B4587-1G B4587-200MG
76219-410EA 224 USD
76219-410 76219-412
2,4-Bis(benzyloxy)-6-chloro-1,3,5-triazine ≥95.0% (by HPLC, titration analysis)
2,4-Bis(benzyloxy)-6-chloro-1,3,5-triazine
Formula: C₁₇H₁₄ClN₃O₂
MW: 327.77 g/mol
CAS Number: 851030-18-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Argentmetric Titration) min. 95.0 %
Melting point 64.0 to 68.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR