N-(4-Chloro-3-cyano-7-ethoxy-6-quinolinyl)acetamide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

C3315-1G C3315-5G
TCC3315-1GEA 70 USD
TCC3315-1G TCC3315-5G
N-(4-Chloro-3-cyano-7-ethoxy-6-quinolinyl)acetamide ≥98.0% (by HPLC, total nitrogen)
N-(4-Chloro-3-cyano-7-ethoxy-6-quinolinyl)acetamide
Formula: C₁₄H₁₂ClN₃O₂
MW: 289.72 g/mol
Melting Pt: 250°C
MDL Number: MFCD09263704
CAS Number: 848133-76-6

Order Now

Specification Test Results

Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR