9-Bromo-10-(9-phenanthryl)anthracene ≥98.0% (by HPLC)

Supplier: TCI America

B4572-1G B4572-200MG
TCB4572-1GEA 465.5 USD
TCB4572-1G TCB4572-200MG
9-Bromo-10-(9-phenanthryl)anthracene ≥98.0% (by HPLC)
9-Bromo-10-(9-phenanthryl)anthracene
Formula: C₂₈H₁₇Br
MDL Number: MFCD27939628
CAS Number: 845457-53-6

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 95.0 %
Melting point 231.0 to 236.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR