3-Ethyl 4-Methyl 2-Aminothiophene-3,4-dicarboxylate ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-Aminothiophene-3,4-dicarboxylic acid 3-Ethyl 4-Methyl ester, 3-Ethyl 4-methyl 2-aminothiophene-3,4-dicarboxylate

E1418-1G E1418-5G
TCE1418-1GEA 110 USD
TCE1418-1G TCE1418-5G
3-Ethyl 4-Methyl 2-Aminothiophene-3,4-dicarboxylate ≥98.0% (by GC)
3-Ethyl 4-Methyl 2-Aminothiophene-3,4-dicarboxylate
Formula: C₉H₁₁NO₄S
MW: 229.25 g/mol
MDL Number: MFCD11977340
CAS Number: 844502-63-2

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 100.0 to 104.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR