5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)oxindole ≥96.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 5-Oxindoleboronic acid Pinacol ester, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)indolin-2-one, 2-(Oxindole-5-yl)-4,4,5,5-Tetramethyl-1,3,2-dioxaborolane

T3512-1G T3512-5G
TCT3512-1GEA 174 USD
TCT3512-1G TCT3512-5G
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)oxindole ≥96.0% (by GC, titration analysis)
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)oxindole
Formula: C₁₄H₁₈BNO₃
MW: 259.11 g/mol
MDL Number: MFCD08669935
CAS Number: 837392-64-0

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 96.0 %
Melting point 232.0 to 236.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR