1-Methyl-3-(4-sulfobutyl)imidazolium Hydrogen sulfate ≥95.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: 1-butylsulphonic-3-methylimidazolium hydrogensulphate

M3120-25G M3120-5G
TCM3120-25GEA 497 USD
TCM3120-25G TCM3120-5G
1-Methyl-3-(4-sulfobutyl)imidazolium Hydrogen sulfate ≥95.0% (by titrimetric analysis)
1-Methyl-3-(4-sulfobutyl)imidazolium Hydrogen sulfate
Formula: C₈H₁₆N₂O₇S₂
MW: 316.34 g/mol
MDL Number: MFCD12761445
CAS Number: 827320-59-2
UN: 3265
ADR: 8 II

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (Neutralization titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR