4-(trans-4-Propylcyclohexyl)phenol ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
4-(Trans-4-propylcyclohexyl)phenol, 1-Hydroxy-4-(trans-4-propylcyclohexyl)benzene
P1616-100G
P1616-25G
TCP1616-100GEA
278
USD
TCP1616-100G
TCP1616-25G
4-(trans-4-Propylcyclohexyl)phenol ≥98.0% (by GC)
4-(trans-4-Propylcyclohexyl)phenol
Formula:
C₁₅H₂₂O MW: 218.34 g/mol Melting Pt: 138°C |
MDL Number:
MFCD00673751 CAS Number: 81936-33-6 EINECS: 440-740-5 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Mesomorphic range | 136.0 to 142.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...