4-Carboxy-3-nitrophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
Synonyms:
4-Borono-2-nitrobenzoic acid, 3-Nitro-4-carboxyphenylboronic acid, 4-Carboxy-3-nitrobenzeneboronic acid
C3233-1G
C3233-200MG
TCC3233-1GEA
166
USD
TCC3233-1G
TCC3233-200MG
4-Carboxy-3-nitrophenylboronic acid (contains varying amounts of Anhydride)
4-Carboxy-3-nitrophenylboronic acid
Formula:
C₇H₆BNO₆ MW: 210.94 g/mol Melting Pt: 220°C |
MDL Number:
MFCD10696661 CAS Number: 80500-28-3 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | 98.0 to 109.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...