2-Methyl-9,10-di(2-naphthyl)anthracene ≥97.0% (by HPLC)
Supplier: TCI America
M2821-1G
M2821-200MG
TCM2821-1GEA
498
USD
TCM2821-1G
TCM2821-200MG
2-Methyl-9,10-di(2-naphthyl)anthracene ≥97.0% (by HPLC)
2-Methyl-9,10-di(2-naphthyl)anthracene
Formula:
C₃₅H₂₄ MW: 444.58 g/mol |
MDL Number:
MFCD12022447 CAS Number: 804560-00-7 |
Specification Test Results
Appearance | White to Yellow to Green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...