2,2-Bis[4-(4-maleimidophenoxy)phenyl]propane ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: Bisphenol A bis(4-maleimidophenyl ether)

B4807-25G B4807-500G
TCB4807-25GEA 73.86 USD
TCB4807-25G TCB4807-500G
2,2-Bis[4-(4-maleimidophenoxy)phenyl]propane ≥98.0% (by HPLC, total nitrogen)
2,2-Bis[4-(4-maleimidophenoxy)phenyl]propane
Formula: C₃₅H₂₆N₂O₆
MW: 570.60 g/mol
MDL Number: MFCD09908251
CAS Number: 79922-55-7

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 163.0 to 167.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR