1-Methyl-3-propylimidazolium chloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
1-Methyl-3-propylimidazolium Chloride
M3035-25G
M3035-5G
76220-330EA
227
USD
76220-330
76220-332
1-Methyl-3-propylimidazolium chloride ≥98.0% (by HPLC, titration analysis)
1-Methyl-3-propylimidazolium chloride
Formula:
C₇H₁₃ClN₂ MW: 160.65 g/mol |
MDL Number:
MFCD17014728 CAS Number: 79917-89-8 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...