Cyanomethyl Dodecyl trithiocarbonate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

Synonyms: Cyanomethyl dodecyl carbonotrithioate

C3626-1G C3626-5G
TCC3626-1GEA 164 USD
TCC3626-1G TCC3626-5G
Cyanomethyl Dodecyl trithiocarbonate ≥98.0% (by HPLC, total nitrogen)
Cyanomethyl Dodecyl trithiocarbonate
Formula: C₁₅H₂₇NS₃
MW: 317.57 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11045090
CAS Number: 796045-97-1

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 29.0 to 33.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR