Tris(2,2'-bipyridine)cobalt(II) bis(hexafluorophosphate) ≥95.0% (by titrimetric analysis)

Supplier: TCI America
Warning

T3255-1G T3255-5G
TCT3255-1GEA 198 USD
TCT3255-1G TCT3255-5G
Tris(2,2'-bipyridine)cobalt(II) bis(hexafluorophosphate) ≥95.0% (by titrimetric analysis)
Tris(2,2'-bipyridine)cobalt(II) bis(hexafluorophosphate)
Formula: C₃₀H₂₄CoF₁₂N₆P₂
MW: 817.43 g/mol
CAS Number: 79151-78-3

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (Chelometric Titration) min. 95.0 %
Elemental analysis (Nitrogen) 9.70 to 10.80 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR