2-Dicyclohexylphosphino-2',6'-diisopropoxybiphenyl ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: [2',6'-Bis(propan-2-yloxy)-[1,1'-biphenyl]-2-yl]dicyclohexylphosphane, Dicyclohexyl(2',6'-diisopropoxy-[1,1'-biphenyl]-2-yl)phosphine, RuPhos, dicyclohexyl-[2-[2,6-di(propan-2-yloxy)phenyl]phenyl]phosphane

D5037-1G D5037-5G
TCD5037-1GEA 227 USD
TCD5037-1G TCD5037-5G
2-Dicyclohexylphosphino-2',6'-diisopropoxybiphenyl ≥98.0% (by HPLC, titration analysis)
2-Dicyclohexylphosphino-2',6'-diisopropoxybiphenyl
Formula: C₃₀H₄₃O₂P
MW: 466.65 g/mol
MDL Number: MFCD06798294
CAS Number: 787618-22-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 123.0 to 127.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR