4-Iodo-4'-propylbiphenyl ≥98.0% (by GC)
Supplier: TCI America
I1010-1G
I1010-200MG
TCI1010-1GEA
150
USD
TCI1010-1G
TCI1010-200MG
4-Iodo-4'-propylbiphenyl ≥98.0% (by GC)
4-Iodo-4'-propylbiphenyl
Formula:
C₁₅H₁₅I MW: 322.19 g/mol |
MDL Number:
MFCD12923241 CAS Number: 782477-81-0 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 130.0 to 134.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...