2-(2-Bromophenyl)-4,6-diphenyl-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

B5663-1G B5663-200MG
76218-876EA 153 USD
76218-876 76218-878
2-(2-Bromophenyl)-4,6-diphenyl-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
2-(2-Bromophenyl)-4,6-diphenyl-1,3,5-triazine
Formula: C₂₁H₁₄BrN₃
MDL Number: MFCD29047055
CAS Number: 77989-15-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 142.0 to 146.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR