9-Aminoanthracene ≥96.0% (by GC, total nitrogen)

Supplier: TCI America
Warning

Synonyms: 9-Anthramine

A1748-100MG A1748-1G A1748-5G
TCA1748-100MGEA 92 USD
TCA1748-100MG TCA1748-1G TCA1748-5G
9-Aminoanthracene ≥96.0% (by GC, total nitrogen)
9-Aminoanthracene
Formula: C₁₄H₁₁N
MW: 193.25 g/mol
Melting Pt: 170°C
MDL Number: MFCD00452690
CAS Number: 779-03-3

Order Now

Specification Test Results

Appearance Yellow to Brown to Dark green powder to crystal
Purity (GC) min. 96.0 %
Purity (with Total Nitrogen) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR