3,5-Bis(methoxymethyloxy)benzyl alcohol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: [3,5-Bis(methoxymethoxy)phenyl]methanol, (3,5-Bis(methoxymethoxy)phenyl)methanol

B5583-1G B5583-5G
TCB5583-1GEA 127 USD
TCB5583-1G TCB5583-5G
3,5-Bis(methoxymethyloxy)benzyl alcohol ≥98.0% (by GC)
3,5-Bis(methoxymethyloxy)benzyl alcohol
Formula: C₁₁H₁₆O₅
MW: 228.24 g/mol
Boiling Pt: 172°C
MDL Number: MFCD27928465
CAS Number: 76280-60-9

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR