1-(4-Bromophenyl)-2-phenylbenzimidazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B4929-1G B4929-5G
TCB4929-1GEA 52.3 USD
TCB4929-1G TCB4929-5G
1-(4-Bromophenyl)-2-phenylbenzimidazole ≥98.0% (by GC, titration analysis)
1-(4-Bromophenyl)-2-phenylbenzimidazole
Formula: C₁₉H₁₃BrN₂
MDL Number: MFCD09746342
CAS Number: 760212-58-6

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 135.0 to 139.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR