5-Fluororesorcinol ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 5-Fluorobenzene-1,3-diol

F1081-1G F1081-200MG
TCF1081-1GEA 936 USD
TCF1081-1G TCF1081-200MG
5-Fluororesorcinol ≥98.0% (by GC, titration analysis)
5-Fluororesorcinol
Formula: C₆H₅FO₂
MW: 128.10 g/mol
MDL Number: MFCD08457182
CAS Number: 75996-29-1

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 136.0 to 140.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR