1,3-Dibromo-5-n-octylbenzene ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
D5257-1G
D5257-5G
76219-248EA
86
USD
76219-248
76219-250
1,3-Dibromo-5-n-octylbenzene ≥98.0% (by GC)
1,3-Dibromo-5-n-octylbenzene
Formula:
C₁₄H₂₀Br₂ MW: 348.12 g/mol |
CAS Number:
75894-99-4 |
Specification Test Results
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...