Methyl 4-Methylcinnamate ≥98.0% (by GC)
Supplier: TCI America
M2825-1G
M2825-5G
TCM2825-1GEA
65
USD
TCM2825-1G
TCM2825-5G
Methyl 4-Methylcinnamate ≥98.0% (by GC)
Methyl 4-Methylcinnamate
Formula:
C₁₁H₁₂O₂ Boiling Pt: 165°C |
MDL Number:
MFCD00460746 CAS Number: 7560-43-2 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 56.0 to 60.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...