9-(3,5-Dibromophenyl)carbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 9-(3,5-Dibromophenyl)-9H-carbazole

D4978-1G D4978-200MG
TCD4978-1GEA 391 USD
TCD4978-1G TCD4978-200MG
9-(3,5-Dibromophenyl)carbazole ≥98.0% (by GC)
9-(3,5-Dibromophenyl)carbazole
Formula: C₁₈H₁₁Br₂N
MW: 401.10 g/mol
MDL Number: MFCD27939624
CAS Number: 750573-26-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 174.0 to 178.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR