9,9'-(5-Bromo-1,3-phenylene)bis(9H-carbazole) ≥97.0% (by HPLC, total nitrogen)

Supplier: TCI America

B5502-1G B5502-5G
TCB5502-1GEA 68 USD
TCB5502-1G TCB5502-5G
9,9'-(5-Bromo-1,3-phenylene)bis(9H-carbazole) ≥97.0% (by HPLC, total nitrogen)
9,9'-(5-Bromo-1,3-phenylene)bis(9H-carbazole)
Formula: C₃₀H₁₉BrN₂
MDL Number: MFCD28100481
CAS Number: 750573-24-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 224.0 to 228.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR