6-Quinoxalinol ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Q0101-1G Q0101-200MG
TCQ0101-1GEA 443 USD
TCQ0101-1G TCQ0101-200MG
6-Quinoxalinol ≥98.0% (by GC, titration analysis)
6-Quinoxalinol
Formula: C₈H₆N₂O
MDL Number: MFCD07364440
CAS Number: 7467-91-6

Order Now

Specification Test Results

Appearance Yellow to Brown to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR