3,6-Dibromo-9-(4-methoxyphenyl)-9H-carbazole ≥98.0% (by HPLC)

Supplier: TCI America
Warning

D5340-1G D5340-5G
TCD5340-1GEA 175 USD
TCD5340-1G TCD5340-5G
3,6-Dibromo-9-(4-methoxyphenyl)-9H-carbazole ≥98.0% (by HPLC)
3,6-Dibromo-9-(4-methoxyphenyl)-9H-carbazole
Formula: C₁₉H₁₃Br₂NO
MW: 431.13 g/mol
MDL Number: MFCD30721572
CAS Number: 746651-52-5

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 168.0 to 172.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR