3-[[tert-Butyl(dimethyl)silyl]oxy]-1-propanol ≥96.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
3-((tert-Butyldimethylsilyl)oxy)propan-1-ol, 3-[[tert-Butyl(dimethyl)silyl]oxy]propan-1-ol, 3-[(tert-Butyldimethylsilyl)oxy]-1-propanol
B5555-1ML
B5555-5ML
TCB5555-1MLEA
55
USD
TCB5555-1ML
TCB5555-5ML
3-[[tert-Butyl(dimethyl)silyl]oxy]-1-propanol ≥96.0% (by GC)
3-[[tert-Butyl(dimethyl)silyl]oxy]-1-propanol
Formula:
C₉H₂₂O₂Si Flash Pt: 93°C |
MDL Number:
MFCD00187968 CAS Number: 73842-99-6 |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...