4-[(4-Bromophenyl)(p-tolyl)amino]benzaldehyde ≥95.0% (by GC)

Supplier: TCI America

B4922-1G B4922-200MG
TCB4922-1GEA 440 USD
TCB4922-1G TCB4922-200MG
4-[(4-Bromophenyl)(p-tolyl)amino]benzaldehyde ≥95.0% (by GC)
4-[(4-Bromophenyl)(p-tolyl)amino]benzaldehyde
Formula: C₂₀H₁₆BrNO
MDL Number: MFCD16658914
CAS Number: 733744-98-4

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 95.0 %
Melting point 115.0 to 119.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR