Bis(4-fluorophenyl)iodonium trifluoromethanesulfonate ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: Bis(4-fluorophenyl)iodonium triflate, Bis(4-fluorophenyl)iodonium Trifluoromethanesulphonate

B5276-1G B5276-5G
TCB5276-1GEA 130.43 USD
TCB5276-1G TCB5276-5G
Bis(4-fluorophenyl)iodonium trifluoromethanesulfonate ≥97.0% (by HPLC, titration analysis)
Bis(4-fluorophenyl)iodonium trifluoromethanesulfonate
Formula: C₁₃H₈F₅IO₃S
MW: 466.16 g/mol
MDL Number: MFCD21608479
CAS Number: 732306-64-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Ion exchange titration) min. 97.0 %
Melting point 168.0 to 172.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR