3-Bromo-9-(p-tolyl)-9H-carbazole ≥97.0% (by GC)

Supplier: TCI America
Warning

B5052-1G B5052-5G
TCB5052-1GEA 96 USD
TCB5052-1G TCB5052-5G
3-Bromo-9-(p-tolyl)-9H-carbazole ≥97.0% (by GC)
3-Bromo-9-(p-tolyl)-9H-carbazole
Formula: C₁₉H₁₄BrN
MDL Number: MFCD16658912
CAS Number: 731016-44-7

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 156.0 to 160.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR