N-(Trifluoromethylthio)phthalimide ≥98.0% (by GC)

Supplier: TCI America
Warning

T3143-1G T3143-5G
TCT3143-1GEA 234 USD
TCT3143-1G TCT3143-5G
N-(Trifluoromethylthio)phthalimide ≥98.0% (by GC)
N-(Trifluoromethylthio)phthalimide
Formula: C₉H₄F₃NO₂S
MDL Number: MFCD18449627
CAS Number: 719-98-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 111.0 to 115.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR