Diethyl [(Tetrahydropyran-2-yloxy)methyl]phosphonate ≥96.0% (by GC)

Supplier: TCI America

D4611-1G D4611-200MG
TCD4611-1GEA 276 USD
TCD4611-1G TCD4611-200MG
Diethyl [(Tetrahydropyran-2-yloxy)methyl]phosphonate ≥96.0% (by GC)
Diethyl [(Tetrahydropyran-2-yloxy)methyl]phosphonate
Formula: C₁₀H₂₁O₅P
Boiling Pt: 95°C
MDL Number: MFCD09263515
CAS Number: 71885-51-3

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR