Methoxy(dimethyl)octadecylsilane ≥90.0% (by GC)
Supplier: TCI America
|
Warning
|
M3011-25ML
M3011-5ML
TCM3011-25MLEA
221
USD
TCM3011-25ML
TCM3011-5ML
Methoxy(dimethyl)octadecylsilane ≥90.0% (by GC)
Methoxy(dimethyl)octadecylsilane
Formula:
C₂₁H₄₆OSi |
MDL Number:
MFCD00042920 CAS Number: 71808-65-6 EINECS: 276-039-4 |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 90.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...