9,9-Dimethyl-10-phenyl-9,10-dihydroacridine ≥98.0% (by GC)

Supplier: TCI America

D5354-1G D5354-200MG
TCD5354-1GEA 393 USD
TCD5354-1G TCD5354-200MG
9,9-Dimethyl-10-phenyl-9,10-dihydroacridine ≥98.0% (by GC)
9,9-Dimethyl-10-phenyl-9,10-dihydroacridine
Formula: C₂₁H₁₉N
MDL Number: MFCD01851187
CAS Number: 717880-39-2

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 117.0 to 121.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR