L-Chicoric acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Chicoric acid, (2R,3R)-2,3-Bis[[(E)-3-(3,4-dihydroxyphenyl)acryloyl]oxy]succinic acid

C3450-10MG C3450-50MG
TCC3450-10MGEA 149 USD
TCC3450-10MG TCC3450-50MG
L-Chicoric acid ≥98.0% (by titrimetric analysis)
L-Chicoric acid
Formula: C₂₂H₁₈O₁₂
MW: 474.37 g/mol
Melting Pt: 206°C
Storage Temperature: Refrigerator
MDL Number: MFCD22683653
CAS Number: 70831-56-0

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -360 to -400 deg (C=1, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR