5-Azoniaspiro[4.4]nonane bis(trifluoromethanesulfonyl)imide ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
A3101-1G
A3101-5G
TCA3101-1GEA
96
USD
TCA3101-1G
TCA3101-5G
5-Azoniaspiro[4.4]nonane bis(trifluoromethanesulfonyl)imide ≥98.0% (by titrimetric analysis)
5-Azoniaspiro[4.4]nonane bis(trifluoromethanesulfonyl)imide
Formula:
C₁₀H₁₆F₆N₂O₄S₂ |
CAS Number:
706759-44-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Ion exchange titration) | min. 98.0 % |
Melting point | 112.0 to 116.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...