1,3,5-Tri(1-naphthyl)benzene ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 1,3,5-Tri(naphthalen-1-yl)benzene

T3193-1G T3193-200MG
TCT3193-1GEA 459 USD
TCT3193-1G TCT3193-200MG
1,3,5-Tri(1-naphthyl)benzene ≥98.0% (by HPLC)
1,3,5-Tri(1-naphthyl)benzene
Formula: C₃₆H₂₄
MW: 456.59 g/mol
MDL Number: MFCD28975119
CAS Number: 7059-70-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 184.0 to 188.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR