2,2,3,3-Tetrafluoro-3-[[1,1,1,2,3,3-hexafluoro-3-[(1,2,2-trifluorovinyl)oxy]propan-2-yl]oxy]propionitrile ≥93.0% (by GC)

Supplier: TCI America

T3493-25G T3493-5G
76220-664EA 702 USD
76220-664 76220-666
2,2,3,3-Tetrafluoro-3-[[1,1,1,2,3,3-hexafluoro-3-[(1,2,2-trifluorovinyl)oxy]propan-2-yl]oxy]propionitrile ≥93.0% (by GC)
2,2,3,3-Tetrafluoro-3-[[1,1,1,2,3,3-hexafluoro-3-[(1,2,2-trifluorovinyl)oxy]propan-2-yl]oxy]propionitrile
Formula: C₈F₁₃NO₂
MW: 389.07 g/mol
Boiling Pt: 102°C
CAS Number: 69804-19-9
EINECS: 274-123-5

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 93.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR