SB-505124 ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 2-[5-(Benzo[d][1,3]dioxol-5-yl)-2-(tert-butyl)-1H-imidazol-4-yl]-6-methylpyridine, 2-(4-(Benzo[d][1,3]dioxol-5-yl)-2-(tert-butyl)-1H-imidazol-5-yl)-6-methylpyridine, 2-[4-(1,3-Benzodioxol-5-yl)-2-(tert-butyl)-1H-imidazol-5-yl]-6-methylpyridine

B6056-25MG B6056-5MG
TCB6056-25MGEA 399 USD
TCB6056-25MG TCB6056-5MG
SB-505124 ≥98.0% (by HPLC)
SB-505124
Formula: C₂₀H₂₁N₃O₂
MDL Number: MFCD11045901
CAS Number: 694433-59-5

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystaline
Purity (HPLC) min. 98.0 area%
Melting point gt. 166.0 to 170.0 deg-C
Elemental analysis (Nitrogen) 11.90 to 13.20 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR