Quinoxaline 1-Oxide ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Q0108-1G Q0108-250MG
TCQ0108-1GEA 217 USD
TCQ0108-1G TCQ0108-250MG
Quinoxaline 1-Oxide ≥97.0% (by GC, titration analysis)
Quinoxaline 1-Oxide
Formula: C₈H₆N₂O
MDL Number: MFCD00054593
CAS Number: 6935-29-1

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 122.0 to 126.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR