Allyl (3-Methylbutoxy)acetate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (3-Methylbutoxy)acetic acid Allyl ester

A3185-25G A3185-5G
TCA3185-25GEA 287 USD
TCA3185-25G TCA3185-5G
Allyl (3-Methylbutoxy)acetate ≥98.0% (by GC)
Allyl (3-Methylbutoxy)acetate
Formula: C₁₀H₁₈O₃
MW: 186.25 g/mol
MDL Number: MFCD03424212
CAS Number: 67634-00-8
EINECS: 266-803-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR