4-Hydroxybenzimidazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

H1547-1G H1547-250MG
TCH1547-1GEA 221 USD
TCH1547-1G TCH1547-250MG
4-Hydroxybenzimidazole ≥98.0% (by GC, titration analysis)
4-Hydroxybenzimidazole
Formula: C₇H₆N₂O
MDL Number: MFCD00963978
CAS Number: 67021-83-4

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 198.0 to 202.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR