1-Ethyl-3-methylimidazolium tricyanomethanide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

E1298-25G E1298-5G
76220-050EA 355 USD
76220-050 76220-052
1-Ethyl-3-methylimidazolium tricyanomethanide ≥98.0% (by HPLC, total nitrogen)
1-Ethyl-3-methylimidazolium tricyanomethanide
Formula: C₁₀H₁₁N₅
MW: 201.23 g/mol
Melting Pt: -10°C
MDL Number: MFCD22460460
CAS Number: 666823-18-3

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR