N-Methyl-N-(trifluoromethylthio)aniline ≥98.0% (by GC)

Supplier: TCI America
Warning

M2595-1G M2595-200MG
TCM2595-1GEA 480 USD
TCM2595-1G TCM2595-200MG
N-Methyl-N-(trifluoromethylthio)aniline ≥98.0% (by GC)
N-Methyl-N-(trifluoromethylthio)aniline
Formula: C₈H₈F₃NS
MW: 207.21 g/mol
Storage Temperature: Refrigerator
CAS Number: 66476-44-6
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR